Learn More
IL-4 Monoclonal Antibody (BVD6-24G2), PE-Cyanine7, eBioscience™, Invitrogen™
Rat Monoclonal Antibody
$138.65 - $418.00
Specifications
Antigen | IL-4 |
---|---|
Clone | BVD6-24G2 |
Applications | Flow Cytometry |
Classification | Monoclonal |
Conjugate | PE-Cyanine7 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
25-704-241
![]() |
Invitrogen™
25704241 |
25 Tests |
Each
for $138.65
Save 25% Reg : |
|
|||||
50-156-07
![]() |
Invitrogen™
25704242 |
100 Tests |
Each for $418.00
|
![]() |
|||||
Description
Description: The BVD6-24G2 antibody reacts with mouse interleukin-4 (IL-4), a 14 kDa cytokine secreted by Th2 cells. Applications Reported: This BVD6-24G2 antibody has been reported for use in intracellular staining followed by flow cytometric analysis. Applications Tested: This BVD6-24G2 antibody has been tested intracellular staining and flow cytometric analysis of restimulated mouse splenocytes cultures. This can be used at less than or equal to 1 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Light sensitivity: This tandem dye is sensitive photo-induced oxidation. Please protect this vial and stained samples from light. Fixation: Samples can be stored in IC Fixation Buffer (cat. 00-8222) (100 μL cell sample + 100 μL IC Fixation Buffer) or 1-step Fix/Lyse Solution (cat. 00-5333) for up to 3 days in the dark at 4°C with minimal impact on brightness and FRET efficiency/compensation. Some generalizations regarding fluorophore performance after fixation can be made, but clone specific performance should be determined empirically. Excitation: 488-561 nm; Emission: 775 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser. Filtration: 0.2 μm post-manufacturing filtered.
Interleukin-4 (IL-4) is a pleiotropic, immune-modulatory cytokine produced by activated T cells, and serves as a ligand for interleukin 4 receptor. IL-4 has been found to have the broadest range of actions. The interleukin 4 receptor also binds to IL13, which may contribute to many overlapping functions of this cytokine and IL13. STAT6, a signal transducer and activator of transcription, has been shown to play an important role in mediating the immune regulatory signal of IL-4. The IL-4 gene, IL-3, IL-5, IL-13, and CSF2 form a cytokine gene cluster on chromosome 5q. In particular, IL-4 is located in close proximity to IL13 on the chromosome. Further, the IL-4 gene, IL13 and IL5 are found to be coordinated by several long-range regulatory elements over a 120 kilobase range on the chromosome. IL-4 delta 2 is a known splice variant of IL-4.Specifications
IL-4 | |
Flow Cytometry | |
PE-Cyanine7 | |
Rat | |
Mouse | |
B cell growth factor 1; B_cell stimulatory factor 1; B-cell growth factor 1; B-cell IgG differentiation factor; B-cell stimulatory factor 1; BCGF1; BCGF-1; binetrakin; BSF1; BSF-1; cytokine; H-IL-4; IGG1 induction factor; IL2; IL4; Il-4; Il4e12; ILN; Interleukin; interleukin 4; interleukin 4 variant 2; interleukin 4 variant IL-4delta2; interleukin 4 variant IL-4delta3; interleukin 4 variant IL-4int2A; interleukin 4 variant IL-4int3A; interleukin 4 variant IL-4int3B; Interleukin4; interleukin-4; interleukin-4 precursor; lymphocyte stimulatory factor 1; MGC79402; M-IL-4; Pitrakinra | |
Il4 | |
Primary | |
4°C, store in dark, DO NOT FREEZE! | |
Il4 |
BVD6-24G2 | |
Monoclonal | |
Liquid | |
RUO | |
P07750 | |
16189 | |
IgG1 κ | |
Affinity chromatography | |
Antibody |
For Research Use Only.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.