Learn More
IL-4 Rat anti-Mouse, Unlabeled, Clone: 11B11, BD
Rat Monoclonal Antibody
$337.50
Specifications
Antigen | IL-4 |
---|---|
Clone | 11B11 |
Concentration | 0.5mg/mL |
Applications | ELISA |
Classification | Monoclonal |
Description
The 11B11 antibody reacts with mouse interleukin-4 (IL-4). The immunogen used to generate the 11B11 hybridoma was partially purified mouse IL-4 from PMA-stimulated EL-4 supernatant. This is a neutralizing antibody.
Host Species: Rat
Clone: 11B11
Isotype: IgG1
Species Reactivity: Mouse
Immunogen: Partially Purified Mouse IL-4
Blocking Assay, Intracellular Staining
Specifications
IL-4 | |
0.5mg/mL | |
Monoclonal | |
Rat | |
Immunology | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
IgG1 | |
Affinity Purified |
11B11 | |
ELISA | |
Unconjugated | |
RUO | |
Mouse | |
Partially Purified Mouse IL-4 | |
Primary | |
Store undiluted at 4°C |
For Research Use Only.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.