Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IL-4R alpha Antibody (209), Alexa Fluor™ 700, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP289472AF700
Description
IL-4R alpha Monoclonal specifically detects IL-4R alpha in Human samples. It is validated for Flow Cytometry.Specifications
IL-4R alpha | |
Monoclonal | |
Alexa Fluor 700 | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Flow Cytometry | |
209 | |
Flow Cytometry | |
CD124 antigen, IL-4 receptor subunit alpha, IL-4R subunit alpha, IL-4RA, IL4RACD124, IL-4R-alpha, interleukin 4 receptor, interleukin-4 receptor alpha chain, interleukin-4 receptor subunit alpha | |
This antibody was obtained from a rabbit immunized with purified, recombinant Human IL-4R alpha (NP_000409.1; Met1-His232). | |
0.1 mL | |
Asthma, B Cell Development and Differentiation Markers, Cancer, Immunology, Myeloid derived Suppressor Cell | |
3566 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction