Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IL-4R alpha Antibody (25463), DyLight 594, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB230M
Description
IL-4R alpha Monoclonal antibody specifically detects IL-4R alpha in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Neutralization, Immunocytochemistry, CyTOFSpecifications
IL-4R alpha | |
Monoclonal | |
DyLight 594 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified from ascites | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot, Flow Cytometry, Immunohistochemistry, Neutralization, Immunocytochemistry, CyTOF | |
25463 | |
Western Blot, Flow Cytometry, Immunohistochemistry, Neutralization, Immunocytochemistry, CyTOF-ready | |
CD124 antigen, IL-4 receptor subunit alpha, IL-4R subunit alpha, IL-4RA, IL4RACD124, IL-4R-alpha, interleukin 4 receptor, interleukin-4 receptor alpha chain, interleukin-4 receptor subunit alpha | |
S. frugiperda insect ovarian cell line Sf 21-derived recombinant human IL-4 R, Gly24-His232, Accession # P24394 | |
0.1 mL | |
Asthma, B Cell Development and Differentiation Markers, Cancer, Immunology, Myeloid derived Suppressor Cell | |
3566 | |
Store at 4°C in the dark. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction