Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IL-4R alpha Rabbit anti-Human, Clone: R001, Novus Biologicals™

Rabbit Monoclonal Antibody
$548.00 - $731.50
Specifications
Antigen | IL-4R alpha |
---|---|
Clone | R001 |
Applications | Neutralization |
Classification | Monoclonal |
Conjugate | Unconjugated |
Description
IL-4R alpha Monoclonal specifically detects IL-4R alpha in Human samples. It is validated for Neutralization.Specifications
IL-4R alpha | |
Neutralization | |
Unconjugated | |
Rabbit | |
Asthma, B Cell Development and Differentiation Markers, Cancer, Immunology, Myeloid derived Suppressor Cell | |
0.2 um filtered solution in Histidine and Arginine buffer, 120mM NaCl, 0.02% Tween 80, pH6.0 | |
3566 | |
IgG | |
Protein A purified |
R001 | |
Monoclonal | |
Purified | |
RUO | |
Human | |
CD124 antigen, IL-4 receptor subunit alpha, IL-4R subunit alpha, IL-4RA, IL4RACD124, IL-4R-alpha, interleukin 4 receptor, interleukin-4 receptor alpha chain, interleukin-4 receptor subunit alpha | |
Recombinant Human IL-4R alpha Protein | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title