Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IL-4R Polyclonal Antibody, Bioss

Rabbit Polyclonal Antibody
Supplier: Bioss BS41425R

This item is not returnable.
View return policy
Description
IL-4R Polyclonal antibody specifically detects IL-4R in Mouse samples. It is validated for ELISA, Western BlotSpecifications
IL-4R | |
Polyclonal | |
Unconjugated | |
Il4ra | |
582J2.1; CD124; il-4 receptor; IL-4 receptor alpha chain; IL-4 receptor subunit alpha; IL-4-binding protein; IL4-BP; Il4r; IL4R nirs variant 1; IL-4R subunit alpha; IL4RA; IL-4RA; IL-4R-alpha; interleukin 4 receptor; interleukin 4 receptor, alpha; interleukin-4 receptor alpha chain; interleukin-4 receptor subunit alpha; Soluble IL-4 receptor subunit alpha; Soluble interleukin-4 receptor subunit alpha | |
Recombinant mouse IL-4R | |
100 μL | |
Primary | |
Mouse | |
Antibody | |
IgG |
ELISA, Western Blot | |
1 mg/mL | |
TBS with 1% BSA, 50% glycerol and 0.03% ProClin 300 | |
P16382 | |
Rabbit | |
Protein A | |
RUO | |
16190 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction