Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IL-5 Antibody (9906), DyLight 650, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB605W
This item is not returnable.
View return policy
Description
IL-5 Monoclonal antibody specifically detects IL-5 in Human samples. It is validated for Western Blot, Flow CytometrySpecifications
IL-5 | |
Monoclonal | |
DyLight 650 | |
B-cell differentiation factor I, EDF, Eosinophil differentiation factor, IL-5T-cell replacing factor, interleukin 5 (colony-stimulating factor, eosinophil), interleukin-5, TRFB cell differentiation factor I | |
S. frugiperda insect ovarian cell line Sf 21-derived recombinant human IL-5, Ile20-Ser134, Accession # P05113 | |
0.5 mL | |
Asthma, Biologically Active Proteins, Cell Cycle and Replication, Cytokine Research, Immunology | |
3567 | |
Store at 4°C in the dark. | |
IgG1 |
Western Blot, Flow Cytometry | |
9906 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified from ascites | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction