Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-5 Monoclonal Antibody (TRFK4), Biotin

IL-5 Monoclonal Antibody (TRFK-4), Biotin

Supplier:  LIFE TECHNOLOGIES AMC0959

Encompass_Preferred

Catalog No. AMC0959

  • $326.65 / Each of 1

    Save 25%

    Reg : $435.00

    The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Only null left
Edge
Add to Cart

Description

Description

TRUSTED_SUSTAINABILITY
Specifications

Specifications

IL-5
Monoclonal
0.5 mg/mL
PBS with 0.1% sodium azide; pH 7.2
P04401
Il5
Recombinant mouse interleukin-5.
100 μg
Primary
Mouse
Antibody
IgG2a
ELISA
TRFK4
Biotin
Il5
B-cell differentiation factor I; B-cell growth factor II; BCGF-II; colony-stimulating factor, eosinophil; cytotoxic T-lymphocyte inducer; EDF; Eosinophil differentiation factor; IL5; IL-5; ILN; Interleukin; Interleukin 5; Interleukin 5 (colony-stimulating factor eosinophil); Interleukin 5 (colony-stimulating factor, eosinophil); Interleukin5; interleukin-5; T-cell replacing factor; TRF
Rat
Purified
RUO
16191
-20°C
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.