Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-5 Rat anti-Human/Mouse, PE, Clone: TRFK5, BD

Catalog No. BDB554395 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.1 mg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB554395 0.1 mg
BDB562049 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB554395 Supplier BD Biosciences Supplier No. 554395
Add to Cart
Add to Cart

Rat Monoclonal Antibody

The TRFK5 antibody reacts with mouse interleukin-5 (IL-5) and cross-reacts with human IL-5. The TRFK5 antibody has been reported to cross react with IL-5 from rhesus monkey. This is a neutralizing antibody.

Host Species: Rat
Clone: TRFK5
Isotype: IgG1 κ
Species Reactivity: Mouse, Human
Immunogen: Mouse Semi-Purified T-Cell Clone Supernatant

Intracellular Staining

TRUSTED_SUSTAINABILITY

Specifications

Antigen IL-5
Applications Flow Cytometry
Classification Monoclonal
Clone TRFK5
Concentration 0.2mg/mL
Conjugate PE
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Rat
Immunogen Mouse Semi-Purified T-Cell Clone Supernatant
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Human, Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.