Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IL-5R alpha/CD125 Antibody (26815), PE/Atto594, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB253PEATT594
Description
IL-5R alpha/CD125 Monoclonal antibody specifically detects IL-5R alpha/CD125 in Human samples. It is validated for Flow CytometrySpecifications
IL-5R alpha/CD125 | |
Monoclonal | |
PE/ATTO 594 | |
PBS | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Primary | |
Human | |
Purified |
Flow Cytometry | |
26815 | |
Flow Cytometry | |
CD125 antigen, CDw125CD125, HSIL5R3, IL-5 receptor subunit alpha, IL5R, IL-5R subunit alpha, IL-5RA, IL-5R-alpha, interleukin 5 receptor type 3, interleukin 5 receptor, alpha, interleukin-5 receptor alpha chain, interleukin-5 receptor alpha subunit, interleukin-5 receptor subunit alpha, MGC26560 | |
S. frugiperda insect ovarian cell line Sf 21-derived recombinant human IL-5 R alpha/CD125, Asp21-Arg335 | |
0.5 mL | |
Immunology | |
3568 | |
Store at 4C in the dark. Do not freeze. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction