Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IL-6 Antibody (076), Alexa Fluor™ 594

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290446AF594
Description
IL-6 Monoclonal specifically detects IL-6 in Mouse samples. It is validated for ELISA.Specifications
IL-6 | |
Monoclonal | |
Alexa Fluor 594 | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Mouse | |
Purified |
ELISA | |
76 | |
ELISA | |
B-cell differentiation factor, B-cell stimulatory factor 2, BSF2, BSF-2, CDF, CTL differentiation factor, HSF, hybridoma growth factor, IFNB2, IFN-beta-2, IL-6, Interferon beta-2, interleukin 6 (interferon, beta 2), interleukin BSF-2, interleukin-6 | |
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse IL-6 (P08505; Phe25-Thr211). | |
0.1 mL | |
Adaptive Immunity, Biologically Active Proteins, Cancer, Cell Cycle and Replication, Cytokine Research, Immunology, Innate Immunity, Neuroscience | |
3569 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction