Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IL-6 Antibody (MQ2-13A5) - Azide and BSA Free, Novus Biologicals™

Rat Monoclonal Antibody
$243.50 - $499.50
Specifications
Antigen | IL-6 |
---|---|
Clone | MQ2-13A5 |
Concentration | 0.5 mg/mL |
Classification | Monoclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP23304701
![]() |
Novus Biologicals
NBP2330470.1MG |
0.1 mg |
Each for $243.50
|
|
|||||
NBP233047
![]() |
Novus Biologicals
NBP233047 |
0.5 mg |
Each for $499.50
|
|
|||||
Description
IL-6 Monoclonal specifically detects IL-6 in Human, Rhesus Macaque samples. It is validated for ELISA.Specifications
IL-6 | |
0.5 mg/mL | |
Unconjugated | |
Rat | |
Adaptive Immunity, Biologically Active Proteins, Cancer, Cell Cycle and Replication, Cytokine Research, Immunology, Innate Immunity, Neuroscience | |
IL6 | |
IgG1 κ | |
Protein A or G purified | |
Human/Rhesus/Cynomolgus IL-6 |
MQ2-13A5 | |
Monoclonal | |
Purified | |
RUO | |
3569 | |
COS-expressed human IL-6 | |
Primary | |
Store at 4C. Do not freeze. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title