Learn More
Invitrogen™ IL-6 Monoclonal Antibody (1936), PE

Description
This antibody does not cross-react with recombinant IL-6 from mouse, rat, or pig.
Specifications
Specifications
| Antigen | IL-6 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | 1936 |
| Conjugate | PE |
| Formulation | Buffered saline solution with BSA and 0.09% sodium azide |
| Gene | Il6 |
| Gene Accession No. | P05231 |
| Gene Alias | B-cell differentiation factor; B-cell hybridoma growth factor; B-cell stimulatory factor 2; BSF2; BSF-2; CDF; CHIL-6; CTL differentiation factor; cytokine; HGF; H-IL-6; HSF; hybridoma growth factor; Ifnb2; IFN-beta-2; Il6; IL-6; ILg6; ILN; interferon beta-2; Interleukin; interleukin 6; Interleukin 6 (interferon, beta 2); interleukin 6 precursor; interleukin BSF-2; interleukin HP-1; Interleukin6; Interleukin-6; interleukin-6 precursor; interleukin-6 protein; M-IL-6; prointerleukin 6; R-IL-6 |
| Gene Symbols | Il6 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.