Learn More
Invitrogen™ IL-6 Monoclonal Antibody (MP5 20F3)
Description
IL-6 Monoclonal Antibody for Western Blot, IHC, Flow, ELISA, Neu
Specifications
Specifications
| Antigen | IL-6 |
| Applications | ELISA, Flow Cytometry, Immunohistochemistry, Neutralization, Western Blot |
| Classification | Monoclonal |
| Clone | MP5 20F3 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative; pH 7.2 |
| Gene | Il6 |
| Gene Accession No. | P08505 |
| Gene Alias | B-cell differentiation factor; B-cell hybridoma growth factor; B-cell stimulatory factor 2; BSF2; BSF-2; CDF; CHIL-6; CTL differentiation factor; cytokine; HGF; H-IL-6; HSF; hybridoma growth factor; Ifnb2; IFN-beta-2; Il6; IL-6; ILg6; ILN; interferon beta-2; Interleukin; interleukin 6; Interleukin 6 (interferon, beta 2); interleukin 6 precursor; interleukin BSF-2; interleukin HP-1; Interleukin6; Interleukin-6; interleukin-6 precursor; interleukin-6 protein; M-IL-6; prointerleukin 6; R-IL-6 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.