Learn More
Invitrogen™ IL-6 Monoclonal Antibody (MP5-20F3), eFluor™ 450, eBioscience™, Invitrogen™
Description
Description: The MP5-20F3 antibody reacts with mouse interleukin-6 (IL-6). The MP5-20F3 antibody is a neutralizing antibody. Mouse IL-6 (also known as B Cell Stimulating Factor, BCSF, and BSF-2) is a 21-28 kDa factor produced by a variety of cell types, including monocytes, fibroblasts and endothelial cells. Upon stimulation, macrophages, T, B, mast, and glial cells, eosinophils, keratinocytes and granulocytes also secrete IL-6. This pleiotrophic cytokine influences the antigen-specific immune response, inflammatory response, and the development of neurons, and is a major mediator of the acute phase reaction. Applications Reported: This MP5-20F3 antibody has been reported for use in intracellular staining followed by flow cytometric analysis. Applications Tested: This MP5-20F3 antibody has been tested by intracellular staining and flow cytometric analysis of stimulated mouse thioglycolate-elicited peritoneal exudate cells using the Intracellular Fixation & Permeabilization Buffer Set (cat. 88-8824) and protocol. Please refer to Best Protocols: Protocol A: Two step protocol for (cytoplasmic) intracellular proteins located under the Resources Tab online. This can be used at less than or equal to 0.5 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test.
Specifications
Specifications
| Antigen | IL-6 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | MP5-20F3 |
| Concentration | 0.2 mg/mL |
| Conjugate | eFluor 450 |
| Formulation | PBS with 0.09% sodium azide; pH 7.2 |
| Gene | Il6 |
| Gene Accession No. | P08505 |
| Gene Alias | B-cell differentiation factor; B-cell hybridoma growth factor; B-cell stimulatory factor 2; BSF2; BSF-2; CDF; CHIL-6; CTL differentiation factor; cytokine; HGF; H-IL-6; HSF; hybridoma growth factor; Ifnb2; IFN-beta-2; Il6; IL-6; ILg6; ILN; interferon beta-2; Interleukin; interleukin 6; Interleukin 6 (interferon, beta 2); interleukin 6 precursor; interleukin BSF-2; interleukin HP-1; Interleukin6; Interleukin-6; interleukin-6 precursor; interleukin-6 protein; M-IL-6; prointerleukin 6; R-IL-6 |
| Show More |
Frequently Asked Questions (FAQs)
Our options will depend on the samples you are analyzing.
If cell viability is not critical, you can store your stained samples at 4 degrees C or on ice overnight in the dark and analyze the following day.
For samples stained with eFluor organic fluorochromes, we recommend that cells be suspended in 100 uL of Flow Cytometry Staining Buffer (Cat. No. 00-4222) and 100 uL of eBioscience IC Fixation Buffer (Cat. No. 00-8222); samples can be incubated for up to 3 days at 4 degrees C in the dark. Alternatively, the 1-step Fix/Lyse Solution (Cat. No. 00-5333) can be used. This is a great option when working with whole blood but also works for other cell types.
The eFluor Organic fluorochromes and eVolve QDots can be used with flow staining buffers containing PBS and protein.
As with other fluorochromes, we recommend minimal exposure to light to maintain optimal signal.
For Research Use Only.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.