Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ IL-6 Monoclonal Antibody (MP5-20F3), eFluor™ 450, eBioscience™, Invitrogen™
GREENER_CHOICE

Catalog No. 501124199
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-112-4199 100 μg
50-112-4198 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-112-4199 Supplier Invitrogen™ Supplier No. 48706182
Add to Cart
Edge
Add to Cart

Rat Monoclonal Antibody

Description: The MP5-20F3 antibody reacts with mouse interleukin-6 (IL-6). The MP5-20F3 antibody is a neutralizing antibody. Mouse IL-6 (also known as B Cell Stimulating Factor, BCSF, and BSF-2) is a 21-28 kDa factor produced by a variety of cell types, including monocytes, fibroblasts and endothelial cells. Upon stimulation, macrophages, T, B, mast, and glial cells, eosinophils, keratinocytes and granulocytes also secrete IL-6. This pleiotrophic cytokine influences the antigen-specific immune response, inflammatory response, and the development of neurons, and is a major mediator of the acute phase reaction. Applications Reported: This MP5-20F3 antibody has been reported for use in intracellular staining followed by flow cytometric analysis. Applications Tested: This MP5-20F3 antibody has been tested by intracellular staining and flow cytometric analysis of stimulated mouse thioglycolate-elicited peritoneal exudate cells using the Intracellular Fixation & Permeabilization Buffer Set (cat. 88-8824) and protocol. Please refer to Best Protocols: Protocol A: Two step protocol for (cytoplasmic) intracellular proteins located under the Resources Tab online. This can be used at less than or equal to 0.5 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test.

IL-6 is a gene that encodes a cytokine that plays a role in inflammation and the maturation of B cells. The protein produced by this gene is an endogenous pyrogen that can induce fever in people with autoimmune diseases or infections. The gene is primarily expressed at sites of acute and chronic inflammation, where it is secreted into the serum and induces a transcriptional inflammatory response through interleukin 6 receptor, alpha. The functioning of this gene is implicated in a wide variety of inflammation-associated disease states, including suspectibility to diabetes mellitus and systemic juvenile rheumatoid arthritis. Elevated levels of the encoded protein have been found in virus infections, including COVID-19 (disease caused by SARS-CoV-2). Diseases associated with IL6 include Kaposi Sarcoma and Rheumatoid Arthritis, Systemic Juvenile.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IL-6
Applications Flow Cytometry
Classification Monoclonal
Clone MP5-20F3
Concentration 0.2 mg/mL
Conjugate eFluor 450
Formulation PBS with 0.09% sodium azide; pH 7.2
Gene Il6
Gene Accession No. P08505
Gene Alias B-cell differentiation factor; B-cell hybridoma growth factor; B-cell stimulatory factor 2; BSF2; BSF-2; CDF; CHIL-6; CTL differentiation factor; cytokine; HGF; H-IL-6; HSF; hybridoma growth factor; Ifnb2; IFN-beta-2; Il6; IL-6; ILg6; ILN; interferon beta-2; Interleukin; interleukin 6; Interleukin 6 (interferon, beta 2); interleukin 6 precursor; interleukin BSF-2; interleukin HP-1; Interleukin6; Interleukin-6; interleukin-6 precursor; interleukin-6 protein; M-IL-6; prointerleukin 6; R-IL-6
Gene Symbols Il6
Host Species Rat
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 16193
Target Species Mouse
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
I am unable to analyze my cells stained with eFluor Organic Dyes today. What options do I have?

Our options will depend on the samples you are analyzing.

If cell viability is not critical, you can store your stained samples at 4 degrees C or on ice overnight in the dark and analyze the following day.

For samples stained with eFluor organic fluorochromes, we recommend that cells be suspended in 100 uL of Flow Cytometry Staining Buffer (Cat. No. 00-4222) and 100 uL of eBioscience IC Fixation Buffer (Cat. No. 00-8222); samples can be incubated for up to 3 days at 4 degrees C in the dark. Alternatively, the 1-step Fix/Lyse Solution (Cat. No. 00-5333) can be used. This is a great option when working with whole blood but also works for other cell types.

Can the eFluor Organic fluorochromes be used for intracellular staining?

Yes, the eFluor Organic fluorochromes can be used for intracellular staining. The eFluor organic fluorochromes maintain bright signal and require minimal changes in compensation when fixed with eBioscience IC Fixation Buffer (Cat. No. 00-8222-49) and Permeabilization Buffer (Cat. No. 00-8333-56) or 1-step Fix/Lyse Solution (Cat. No. 00-5333-54, 00-5333-57) (as compared to live cells).

What buffers are compatible with the eFluor Organic fluorochromes and eVolve QDots?

The eFluor Organic fluorochromes and eVolve QDots can be used with flow staining buffers containing PBS and protein.

What is the difference between eFluor Organic Dyes and eVolve Qdots?

The eFluor Organic Dyes (eFluor 450, APC-eFluor 780, PerCP-eFluor 710, eFluor 710) are conventional fluorochromes. In contrast, the eVolve line of products are Quantum dots.

Are the eFluor Organic Dyes photo-labile?

As with other fluorochromes, we recommend minimal exposure to light to maintain optimal signal.

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.