Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IL-6 Rat anti-Mouse, PE, Clone: MP5-20F3, BD
Rat Monoclonal Antibody
$122.00 - $303.50
Specifications
Antigen | IL-6 |
---|---|
Clone | MP5-20F3 |
Concentration | 0.2mg/mL |
Applications | Flow Cytometry |
Classification | Monoclonal |
Description
The MP5-20F3 monoclonal antibody specifically binds to mouse interleukin-6 (IL-6). The immunogen used to generate the MP5-20F3 hybridoma was recombinant mouse IL-6.
Specifications
IL-6 | |
0.2mg/mL | |
Monoclonal | |
Rat | |
Mouse | |
Mouse IL-6 Recombinant Protein | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
MP5-20F3 | |
Flow Cytometry | |
PE | |
RUO | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
IgG1 | |
Affinity Purified |
For Research Use Only.
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title