Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IL-6R alpha Antibody (033), mFluor Violet 450 SE, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290512MFV450
Description
IL-6R alpha Monoclonal antibody specifically detects IL-6R alpha in Mouse samples. It is validated for Flow CytometrySpecifications
IL-6R alpha | |
Monoclonal | |
mFluor Violet 450 SE | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
3570 | |
Store at 4C in the dark. | |
IgG |
Flow Cytometry | |
033 | |
Flow Cytometry | |
CD126, gp80, IL6Q, IL6R, IL-6R-1, IL6RA, IL-6RA, IL6RQ, interleukin 6 receptor | |
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse IL-6R alpha (NP_034689.2; Ile44-Glu357). | |
0.1 mL | |
Primary | |
Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction