Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IL-6R alpha Antibody (17506R), PE/Atto594, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB227RPEATT594
Description
IL-6R alpha Monoclonal antibody specifically detects IL-6R alpha in Human samples. It is validated for Flow CytometrySpecifications
IL-6R alpha | |
Monoclonal | |
PE/ATTO 594 | |
PBS | |
Mouse | |
Protein A or G purified from cell culture supernatant | |
RUO | |
3570 | |
Store at 4C in the dark. Do not freeze. | |
IgG1 |
Flow Cytometry | |
17506R | |
Flow Cytometry | |
CD126, gp80, IL6Q, IL6R, IL-6R-1, IL6RA, IL-6RA, IL6RQ, interleukin 6 receptor | |
S. frugiperda insect ovarian cell line Sf 21-derived recombinant human IL-6 R alpha , Leu20-Asp339, Accession # P08887 | |
0.1 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction