Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-6R Beta/CD130/gp130 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501983305
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-3305 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-3305 Supplier Bioss Supplier No. BS1459R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The protein encoded by this gene is a signal transducer shared by many cytokines, including interleukin 6 (IL6), ciliary neurotrophic factor (CNTF), leukemia inhibitory factor (LIF), and oncostatin M (OSM). This protein functions as a part of the cytokine receptor complex. The activation of this protein is dependent upon the binding of cytokines to their receptors. vIL6, a protein related to IL6 and encoded by the Kaposi sarcoma-associated herpesvirus, can bypass the interleukin 6 receptor (IL6R) and directly activate this protein. Knockout studies in mice suggested a critical role of the gene encoding this protein in regulating myocyte apoptosis. Alternatively spliced transcript variants encoding distinct isoforms have been described.

Specifications

Antigen IL-6R Beta/CD130/gp130
Applications Flow Cytometry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene IL6ST
Gene Accession No. P40189
Gene Alias 5133400A03Rik; AA389424; Ac1055; BB405851; CD130; CD130 antigen; CDw130; D13Ertd699e; EGK_16499; gp130; gp130 of the rheumatoid arthritis antigenic peptide-bearing soluble form; gp130, oncostatin M receptor; GP130-RAPS; IL-6; IL-6 receptor subunit beta; IL-6R subunit beta; Il-6rb; IL-6R-beta; IL6R-beta; IL6ST; interleukin 6 signal transducer; interleukin 6 signal transducer (gp130, oncostatin M receptor); interleukin 6 signal transducer receptor; interleukin receptor beta chain; Interleukin-6 receptor subunit beta; Interleukin-6 signal transducer; Membrane glycoprotein 130; membrane glycoprotein gp130; Oncostatin-M receptor subunit alpha
Gene Symbols IL6ST
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from mouse IL6R beta.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3572
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.