Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IL-7 Antibody (040), mFluor Violet 450 SE, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290485MFV450
Description
IL-7 Monoclonal antibody specifically detects IL-7 in Mouse samples. It is validated for ELISASpecifications
IL-7 | |
Monoclonal | |
mFluor Violet 450 SE | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Mouse | |
Purified |
ELISA | |
040 | |
ELISA | |
IL-7interleukin-7, interleukin 7 | |
Rabbit immunized with purified, recombinant Mouse IL-7 (Accession#: NP_032397.1, Glu26-Ile154) | |
0.1 mL | |
Biologically Active Proteins, Cell Cycle and Replication, Cytokine Research, Hematopoietic Stem Cell Markers, Immunology, Innate Immunity, Stem Cell Markers | |
3574 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction