Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IL-7 Antibody (MM0410-12T6) - Azide and BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
$206.50
Specifications
Antigen | IL-7 |
---|---|
Clone | MM0410-12T6 |
Concentration | LYOPH |
Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Blocking Assay |
Classification | Monoclonal |
Description
IL-7 Monoclonal specifically detects IL-7 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Block/Neutralize.Specifications
IL-7 | |
LYOPH | |
Monoclonal | |
Purified | |
RUO | |
P13232 | |
3574 | |
IgG1 | |
Protein G purified |
MM0410-12T6 | |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Blocking Assay | |
Unconjugated | |
Mouse | |
Human | |
IL-7interleukin-7, interleukin 7 | |
IL7 | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title