Learn More
Invitrogen™ IL-7 Polyclonal Antibody

Description
Positive Control: SHSY5Y cell lysate, HepG2 cell lysate, PANC-1, SH-SY5Y, SiHa, human tonsil tissue, human spleen tissue. Subcellular Location: Secreted.
Specifications
Specifications
| Antigen | IL-7 |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | TBS with 0.2% BSA, 50% glycerol and 0.05% sodium azide; pH 7.4 |
| Gene | Il7 |
| Gene Accession No. | P13232, P16871 |
| Gene Alias | A630026I06Rik; H-IL-7; hlb368; IL7; Il-7; ILN; Interleukin; interleukin 7; Interleukin7; interleukin-7; M-IL-7 |
| Gene Symbols | Il7, IL7r |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.