Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ IL-7 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5120457
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5120457 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5120457 Supplier Invitrogen™ Supplier No. PA5120457
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive test controls include: THP-1 . The target is usually found in the following locations: Secreted. Immunogen sequence: DCDIEGKDGK QYESVLMVSI DQLLDSMKEI GSNCLNNEFN FFKRHICDAN KEGMFLFRAA RKLRQFLKMN STGDFDLHLL KVSEGTTILL NCTGQVKGRK PAALGEAQPT KSLEENKSLK EQKKLNDLCF LKRLLQEIKT CWNKILMGTK EH.

Interleukin 7 (IL-7) is a hematopoietic cytokine that is constitutively produced by stromal cells from the bone marrow and thymus. IL-7 signals through the IL-7R to promote the differentiation of hematopoietic stem cells to lymphoid precursor cells, which give rise to T cells, B cells, and natural killer cells.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IL-7
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 2.36 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3
Gene Il7
Gene Accession No. P10168, P13232, P16871, P16872, P56478
Gene Alias A630026I06Rik; H-IL-7; hlb368; IL7; Il-7; ILN; Interleukin; interleukin 7; Interleukin7; interleukin-7; M-IL-7
Gene Symbols Il7, IL7r
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 26-177 of human IL7 (NP_0008711).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 16196, 16197, 25647, 294797, 3574, 3575
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.