Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IL-7R alpha/CD127 Antibody (22F3D9) - Azide and BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP280812
Description
IL-7R alpha/CD127 Monoclonal specifically detects IL-7R alpha/CD127 in Human samples. It is validated for Western Blot, Flow Cytometry, CyTOF-ready.Specifications
IL-7R alpha/CD127 | |
Monoclonal | |
1 mg/mL | |
Western Blot : 2-5 ug/ml, Flow Cytometry 1:100. Use reported by customer review, CyTOF-ready | |
CD127 antigen, CD127ILRA, CDW127, IL-7 receptor subunit alpha, IL-7R subunit alpha, IL7RA, IL-7RA, IL-7R-alpha, interleukin 7 receptor, interleukin 7 receptor alpha chain, interleukin 7 receptor isoform H5-6, interleukin-7 receptor subunit alpha | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot, Flow Cytometry, CyTOF | |
22F3D9 | |
Unconjugated | |
PBS | |
IL7R | |
Amino acids 141-237 of human CD127 were used as the immunogen for the antibody. | |
0.1 mg | |
Apoptosis, Cytokine Research, Immunology, Innate Immunity, Phospho Specific | |
3575 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
IgG2b λ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction