Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IL-7R alpha/CD127 Antibody (717519), mFluor Violet 610 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB8484MFV610
Description
IL-7R alpha/CD127 Monoclonal antibody specifically detects IL-7R alpha/CD127 in Rat samples. It is validated for Flow Cytometry, CyTOFSpecifications
| IL-7R alpha/CD127 | |
| Monoclonal | |
| mFluor Violet 610 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| 0.1 mL | |
| Apoptosis, Cytokine Research, Immunology, Innate Immunity, Phospho Specific | |
| 3575 | |
| Store at 4C in the dark. |
| Flow Cytometry, CyTOF | |
| 717519 | |
| Flow Cytometry, CyTOF-ready | |
| CD127 antigen, CD127ILRA, CDW127, IL-7 receptor subunit alpha, IL-7R subunit alpha, IL7RA, IL-7RA, IL-7R-alpha, interleukin 7 receptor, interleukin 7 receptor alpha chain, interleukin 7 receptor isoform H5-6, interleukin-7 receptor subunit alpha | |
| Mouse myeloma cell line NS0-derived recombinant rat IL-7 R alpha/CD127, Glu21-Asp239, Accession # NP_001099888 | |
| RUO | |
| Primary | |
| Rat | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction