Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IL-7R alpha/CD127 Antibody (73N8F2), Novus Biologicals™

Rat Monoclonal Antibody
$206.50 - $415.50
Specifications
| Antigen | IL-7R alpha/CD127 |
|---|---|
| Clone | 73N8F2 |
| Concentration | 1.0 mg/ml |
| Dilution | Western Blot 2-5 ug/ml, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 5 ug/ml |
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP225249SS
![]() |
Novus Biologicals
NBP225249SS |
0.025 mg |
Each for $206.50
|
|
|||||
NBP225249
![]() |
Novus Biologicals
NBP225249 |
0.1 mg |
Each for $415.50
|
|
|||||
Description
IL-7R alpha/CD127 Monoclonal specifically detects IL-7R alpha/CD127 in Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| IL-7R alpha/CD127 | |
| 1.0 mg/ml | |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rat | |
| Apoptosis, Cytokine Research, Immunology, Innate Immunity, Phospho Specific | |
| NP_032398 | |
| 3575 | |
| Amino acids 145-235 of mouse IL-7R CD127 were used as the immunogen for the antibody. | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| 73N8F2 | |
| Western Blot 2-5 ug/ml, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 5 ug/ml | |
| Monoclonal | |
| Purified | |
| RUO | |
| Mouse | |
| CD127 antigen, CD127ILRA, CDW127, IL-7 receptor subunit alpha, IL-7R subunit alpha, IL7RA, IL-7RA, IL-7R-alpha, interleukin 7 receptor, interleukin 7 receptor alpha chain, interleukin 7 receptor isoform H5-6, interleukin-7 receptor subunit alpha | |
| IL7R | |
| IgG2a κ | |
| Protein G purified | |
| This product reacts with Mouse. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title