Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IL-7R alpha/CD127 Antibody (R34-34), Alexa Fluor™ 647, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals DDX0700A647100
Description
IL-7R alpha/CD127 Monoclonal specifically detects IL-7R alpha/CD127 in Human, Canine samples. It is validated for Flow Cytometry, Product Image.Specifications
IL-7R alpha/CD127 | |
Monoclonal | |
0.5mg/mL | |
Flow Cytometry 1:10-1:1000 | |
CD127 antigen, CD127ILRA, CDW127, IL-7 receptor subunit alpha, IL-7R subunit alpha, IL7RA, IL-7RA, IL-7R-alpha, interleukin 7 receptor, interleukin 7 receptor alpha chain, interleukin 7 receptor isoform H5-6, interleukin-7 receptor subunit alpha | |
Mouse | |
Ion exchange chromatography | |
RUO | |
Primary | |
Human, Canine | |
IgG1 |
Flow Cytometry | |
R34-34 | |
Alexa Fluor 647 | |
NP_002176 | |
IL7R | |
IL7R Alpha/CD127 was used as immunogen. | |
0.1 mg | |
Apoptosis, Cytokine Research, Immunology, Innate Immunity, Phospho Specific | |
3575 | |
Store at -20C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction