Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IL-7R alpha/CD127 Antibody (YIL7R323), DyLight 680, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP281081FR
Description
IL-7R alpha/CD127 Monoclonal specifically detects IL-7R alpha/CD127 in Human samples. It is validated for Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence.Specifications
IL-7R alpha/CD127 | |
Monoclonal | |
DyLight 680 | |
50mM Sodium Borate | |
IL7R | |
Recombinant monoclonal antibody to IL-7R alpha/CD127r. | |
0.1 mL | |
Apoptosis, Cytokine Research, Immunology, Innate Immunity, Phospho Specific | |
3575 | |
Human | |
Purified |
Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence | |
YIL7R323 | |
Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence | |
CD127 antigen, CD127ILRA, CDW127, IL-7 receptor subunit alpha, IL-7R subunit alpha, IL7RA, IL-7RA, IL-7R-alpha, interleukin 7 receptor, interleukin 7 receptor alpha chain, interleukin 7 receptor isoform H5-6, interleukin-7 receptor subunit alpha | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
This antibody binds to an epitope of human IL-7R alpha/CD127 distinct to that bound by Ab00531. | |
Store at 4°C in the dark. | |
IgG κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction