Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IL-9 Antibody (103), DyLight 488

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP289999G
Description
IL-9 Monoclonal specifically detects IL-9 in Human samples. It is validated for ELISA.Specifications
| IL-9 | |
| Monoclonal | |
| DyLight 488 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| ELISA | |
| 103 | |
| ELISA | |
| Cytokine P40, HP40, IL-9homolog of mouse T cell and mast cell growth factor 40, interleukin 9, interleukin-9, P40, p40 cytokine, p40 T-cell and mast cell growth factor, T-cell growth factor P40 | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Human IL-9 (NP_000581.1; Gln19-Ile144). | |
| 0.1 mL | |
| Asthma, Biologically Active Proteins, Cell Cycle and Replication, Cytokine Research, Immunology, Innate Immunity | |
| 3578 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction