Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IL-9 Antibody (222604), Janelia Fluor™ 549, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals FAB409I
Description
IL-9 Monoclonal antibody specifically detects IL-9 in Mouse samples. It is validated for Western Blot, Flow Cytometry, CyTOFSpecifications
IL-9 | |
Monoclonal | |
Janelia Fluor 549 | |
50mM Sodium Borate | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Primary | |
Mouse | |
Purified |
Western Blot, Flow Cytometry, CyTOF | |
222604 | |
Western Blot, Intracellular Staining by Flow Cytometry, CyTOF-ready | |
Cytokine P40, HP40, IL-9homolog of mouse T cell and mast cell growth factor 40, interleukin 9, interleukin-9, P40, p40 cytokine, p40 T-cell and mast cell growth factor, T-cell growth factor P40 | |
S. frugiperda insect ovarian cell line Sf 21-derived recombinant mouse IL-9, Gln19-Pro144, Accession # P15247.1 | |
0.5 mL | |
Asthma, Biologically Active Proteins, Cell Cycle and Replication, Cytokine Research, Immunology, Innate Immunity | |
3578 | |
Store at 4C in the dark. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction