Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IL-9 Antibody (D9302C12), DyLight 650, Novus Biologicals™

Armenian Hamster Monoclonal Antibody
Supplier: Novus Biologicals NBP312011C
Description
IL-9 Monoclonal antibody specifically detects IL-9 in Mouse samples. It is validated for Flow CytometrySpecifications
IL-9 | |
Monoclonal | |
DyLight 650 | |
50mM Sodium Borate | |
Armenian Hamster | |
Protein A purified | |
RUO | |
Primary | |
Mouse | |
Purified |
Flow Cytometry | |
D9302C12 | |
Flow Cytometry | |
Cytokine P40, HP40, IL-9homolog of mouse T cell and mast cell growth factor 40, interleukin 9, interleukin-9, P40, p40 cytokine, p40 T-cell and mast cell growth factor, T-cell growth factor P40 | |
This antibody was raised by immunising Armenian hamsters with Baculovirus-expressed, recombinant murine IL-9. | |
0.1 mL | |
Asthma, Biologically Active Proteins, Cell Cycle and Replication, Cytokine Research, Immunology, Innate Immunity | |
3578 | |
Store at 4C in the dark. | |
IgG κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction