Learn More
Invitrogen™ IL-9 Chimeric Recombinant Rabbit Monoclonal Antibody (RM9A4)

Description
Specificity: This antibody is specific for murine IL-9.
Specifications
Specifications
| Antigen | IL-9 Chimeric |
| Applications | ELISA, Flow Cytometry |
| Classification | Recombinant Monoclonal |
| Clone | RM9A4 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.02% ProClin 300 |
| Gene | Il9 |
| Gene Accession No. | P15247 |
| Gene Alias | cytokine P40; homolog of mouse T cell and mast cell growth factor 40; HP40; IL9; IL-9; ILN; Interleukin; interleukin 9; Interleukin9; interleukin-9; P40; p40 cytokine; p40 T-cell and mast cell growth factor; T-cell growth factor P40 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.