Learn More
Invitrogen™ IL-9 Monoclonal Antibody (MH9A4), eFluor™ 660, eBioscience™
Mouse Monoclonal Antibody
Supplier: Invitrogen™ 50709742

Description
Description: The monoclonal antibody MH9A4 recognizes interleukin-9 (IL-9), a proinflammatory cytokine historically believed to be involved in type 2 immune responses. Recent evidence suggests IL-9 may instead be secreted by a distinct T-helper lineage called Th9. These cells can be derived from Th2 cells with TGF beta or differentiated directly from naive CD4+ T cells with TGF beta and IL-4. IL-9 is a member of the common cytokine receptor gamma chain-dependent family of cytokines which also includes IL-2, IL-4, IL-7, IL-15 and IL-21. Its pleiotropic effects on Th2 lymphocytes, B lymphocytes, mast cells, eosinophils and gut and airway epithelial cells have implicated IL-9 in asthma and other allergy-related disorders. Staining with the MH9A4 antibody is not blocked by the MH9D1 antibody, suggesting that the two antibodies bind to unique epitopes. Applications Reported: This MH9A4 antibody has been reported for use in intracellular staining followed by flow cytometric analysis. Applications Tested: This MH9A4 antibody has been pre-titrated and tested by intracellular staining and flow cytometric analysis of stimulated human peripheral blood cells. This can be used at 5 μL (0.06 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. eFluor® 660 is a replacement for Alexa Fluor® 647. eFluor® 660 em...
Interleukin 9 (IL-9) is produced by activated T helper 2 (Th2) lymphocytes. IL-9 regulates many cell types involved in classical Th2-associated asthma responses, including B and T lymphocytes, mast cells, eosinophils, and epithelial cells. The mouse IL-9 cDNA encodes a 144 amino acid precursor protein with an 18 amino acid signal peptide that is cleaved to form the mature cysteine-rich protein with a molecular mass of 14 kDa. Native mouse IL-9 is heavily glycosylated. Recently, TGF beta has been reported to play an important role in the differentiation of IL-17-producing T helper cells and inducible regulatory T cells by reprogramming Th2 cells to secrete IL-9, thereby driving the differentiation of Th9 cells directly.
Specifications
IL-9 | |
Monoclonal | |
5 μL/Test | |
PBS with BSA and 0.09% sodium azide; pH 7.2 | |
P15248 | |
Il9 | |
Affinity chromatography | |
RUO | |
3578 | |
4°C, store in dark, DO NOT FREEZE! | |
Liquid |
Flow Cytometry | |
MH9A4 | |
eFluor 660 | |
Il9 | |
cytokine P40; homolog of mouse T cell and mast cell growth factor 40; HP40; IL9; IL-9; ILN; Interleukin; interleukin 9; Interleukin9; interleukin-9; P40; p40 cytokine; p40 T-cell and mast cell growth factor; T-cell growth factor P40 | |
Mouse | |
100 Tests | |
Primary | |
Human | |
Antibody | |
IgG2b κ |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.