Learn More
IL-9 Polyclonal Antibody, PeproTech®, Invitrogen™
Rabbit Polyclonal Antibody
$250.00
Specifications
Antigen | IL-9 |
---|---|
Concentration | 0.1-1.0 mg/mL |
Applications | ELISA, Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
500P2950UG
![]() |
Invitrogen™
500P2950UG |
50 μg |
Each of 1 for $250.00
|
|
|||||
Description
AA Sequence of recombinant protein: MQGCPTLAGI LDINFLINKM QEDPASKCHC SANVTSCLCL GIPSDNCTRP CFSERLSQMT NTTMQTRYPL IFSRVKKSVE VLKNNKCPYF SCEQPCNQTT AGNALTFLKS LLEIFQKEKM RGMRGKI. Preparation: Produced from sera of rabbits immunized with highly pure Recombinant Human IL-9. Anti-Human IL-9-specific antibody was purified by affinity chromatography employing an immobilized Human IL-9 matrix. Sandwich ELISA: To detect Human IL-9 by sandwich ELISA (using 100 μL/well antibody solution) a concentration of 0.5-2.0 μg/mL of this antibody is required. This antigen affinity purified antibody, in conjunction with PeproTech Biotinylated Anti-Human IL-9 (500-P29Bt) as a detection antibody, allows the detection of at least 0.2-0.4 ng/well of Recombinant Human IL-9. Western Blot: To detect Human IL-9 by Western Blot analysis this antibody can be used at a concentration of 0.1-0.2μg/mL. When used in conjunction with compatible secondary reagents, the detection limit for Recombinant Human IL-9 is 1.5-3.0 ng/lane, under either reducing or non-reducing conditions.
Interleukin 9 (IL-9) is produced by activated T helper 2 (Th2) lymphocytes. IL-9 regulates many cell types involved in classical Th2-associated asthma responses, including B and T lymphocytes, mast cells, eosinophils, and epithelial cells. The mouse IL-9 cDNA encodes a 144 amino acid precursor protein with an 18 amino acid signal peptide that is cleaved to form the mature cysteine-rich protein with a molecular mass of 14 kDa. Native mouse IL-9 is heavily glycosylated. Recently, TGF beta has been reported to play an important role in the differentiation of IL-17-producing T helper cells and inducible regulatory T cells by reprogramming Th2 cells to secrete IL-9, thereby driving the differentiation of Th9 cells directly.Specifications
IL-9 | |
ELISA, Western Blot | |
Unconjugated | |
Rabbit | |
Human | |
P15248 | |
3578 | |
E.coli-derived Recombinant Human IL-9. | |
Antigen affinity chromatography | |
Il9 |
0.1-1.0 mg/mL | |
Polyclonal | |
Lyophilized | |
RUO | |
PBS with no preservative | |
cytokine P40; homolog of mouse T cell and mast cell growth factor 40; HP40; IL9; IL-9; ILN; Interleukin; interleukin 9; Interleukin9; interleukin-9; P40; p40 cytokine; p40 T-cell and mast cell growth factor; T-cell growth factor P40 | |
Il9 | |
Primary | |
-20°C |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.