Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IL-9 Rabbit anti-Mouse, Clone: RM9A4, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP3120230.2MG
Description
IL-9 Monoclonal antibody specifically detects IL-9 in Mouse samples. It is validated for Flow Cytometry, ELISASpecifications
IL-9 | |
Monoclonal | |
1 mg/ml | |
Flow Cytometry, ELISA | |
Cytokine P40, HP40, IL-9homolog of mouse T cell and mast cell growth factor 40, interleukin 9, interleukin-9, P40, p40 cytokine, p40 T-cell and mast cell growth factor, T-cell growth factor P40 | |
This antibody was raised by immunising rats with recombinant full-length murine IL-9. | |
0.2 mg | |
Asthma, Biologically Active Proteins, Cell Cycle and Replication, Cytokine Research, Immunology, Innate Immunity | |
3578 | |
Store at 4C for up to 3 months. For longer storage, aliquot and store at -20C. | |
IgG κ |
Flow Cytometry, ELISA | |
RM9A4 | |
Unconjugated | |
PBS | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction