Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-9 Receptor Alpha (CD129) Mouse anti-Human, BUV395, Clone: AH9R7, BD OptiBuild™

Catalog No. BDB745622 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB745622 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB745622 Supplier BD Biosciences Supplier No. 745622
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

IL-9 Receptor Alpha (CD129) Monoclonal antibody specifically detects IL-9 Receptor Alpha (CD129) in Human samples. It is validated for Flow Cytometry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IL-9 Receptor Alpha (CD129)
Applications Flow Cytometry
Classification Monoclonal
Clone AH9R7
Concentration 0.2 mg/mL
Conjugate Brilliant Ultraviolet 395
Host Species Mouse
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Refrigerate (2 to 8°C)
Form Purified
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.