Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IL-9R Antibody (33423), DyLight 350, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB290D
This item is not returnable.
View return policy
Description
IL-9R Monoclonal antibody specifically detects IL-9R in Human samples. It is validated for Flow CytometrySpecifications
| IL-9R | |
| Monoclonal | |
| DyLight 350 | |
| CD129, CD129 antigen, IL-9 receptor, IL-9R, interleukin 9 receptor, interleukin-9 receptor | |
| S. frugiperda insect ovarian cell line Sf 21-derived recombinant human IL-9 R | |
| 0.5 mL | |
| Cytokine Research | |
| 3581 | |
| Store at 4°C in the dark. | |
| IgG1 |
| Flow Cytometry | |
| 33423 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified from ascites | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction