Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IL-9R Antibody (AH9R2), Alexa Fluor™ 488, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP312033AF488
Description
IL-9R Monoclonal antibody specifically detects IL-9R in Human samples. It is validated for Flow Cytometry, ImmunofluorescenceSpecifications
IL-9R | |
Monoclonal | |
Alexa Fluor 488 | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
3581 | |
Store at 4C in the dark. | |
IgG κ |
Flow Cytometry, Immunofluorescence | |
AH9R2 | |
Flow Cytometry, Immunocytochemistry/Immunofluorescence | |
CD129, CD129 antigen, IL-9 receptor, IL-9R, interleukin 9 receptor, interleukin-9 receptor | |
This antibody was raised by immunising DBA/2 mice with P815 mastocytoma cells transfected with human IL-9R cDNA. | |
0.1 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction