Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IL-9R Antibody (AH9R7), Allophycocyanin, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP236513APC
Description
IL-9R Monoclonal specifically detects IL-9R in Human samples. It is validated for Flow Cytometry.Specifications
IL-9R | |
Monoclonal | |
APC | |
PBS with 0.05% Sodium Azide | |
IL9R | |
Human CD129-transfected cell line | |
0.1 mL | |
Primary | |
The mouse monoclonal antibody AH9R7 recognizes CD129 IL-9R alpha, a 57 kDa type I transmembrane glycoprotein expressed at low levels by lymphocytes, blood cell progenitors, eosinophils, mast cells, epithelial cells, muscle cells and neurons. | |
Store at 4C in the dark. | |
IgG2b κ |
Flow Cytometry | |
AH9R7 | |
Flow Cytometry | |
CD129, CD129 antigen, IL-9 receptor, IL-9R, interleukin 9 receptor, interleukin-9 receptor | |
Mouse | |
Protein A purified | |
RUO | |
3581 | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction