Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IL-9R Antibody (RM0115-12F43), Allophycocyanin, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals NBP122493APC
Description
IL-9R Monoclonal antibody specifically detects IL-9R in Mouse samples. It is validated for Western BlotSpecifications
IL-9R | |
Monoclonal | |
APC | |
PBS | |
Rat | |
Protein A or G purified | |
RUO | |
Primary | |
Mouse | |
Purified |
Western Blot | |
RM0115-12F43 | |
Western Blot | |
CD129, CD129 antigen, IL-9 receptor, IL-9R, interleukin 9 receptor, interleukin-9 receptor | |
Mouse recombinant protein of the extracellular domain of IL-9 R | |
0.1 mL | |
Cytokine Research | |
3581 | |
Store at 4C in the dark. | |
IgG2 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction