Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IL-9R Antibody (RM0115-12F43), PE, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals NBP122493PE
Description
IL-9R Monoclonal antibody specifically detects IL-9R in Mouse samples. It is validated for Western BlotSpecifications
| IL-9R | |
| Monoclonal | |
| PE | |
| PBS | |
| Rat | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
| Western Blot | |
| RM0115-12F43 | |
| Western Blot | |
| CD129, CD129 antigen, IL-9 receptor, IL-9R, interleukin 9 receptor, interleukin-9 receptor | |
| Mouse recombinant protein of the extracellular domain of IL-9 R | |
| 0.1 mL | |
| Cytokine Research | |
| 3581 | |
| Store at 4C in the dark. | |
| IgG2 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction