Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-9R (CD129) Mouse, anti-Human, R718, Clone: AH9R7, BD OptiBuild™

Catalog No. BDB752104 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB752104 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB752104 Supplier BD Biosciences Supplier No. 752104
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The AH9R7 monoclonal antibody specifically binds to the Interleukin-9 Receptor (IL-9R, IL-9Rα) also known as CD129. CD129 is a Type I transmembrane glycoprotein and a Type I Cytokine Receptor (Hemopoietin) Superfamily member. The high-affinity, signaling IL-9 Receptor complex is comprised of CD129 (IL-9 Receptor alpha chain subunit) and CD132 (common γ chain/γc). CD129 is expressed by hematopoietic progenitors, thymocytes, T cells, B cells, macrophages, eosinophils, mast cells, epithelia cells, muscle cells and neurons. IL-9 signals through the IL-9R complex (ie, through JAK, STAT and related signal pathways) to regulate the growth, proliferation, differentiation and survival of myeloid and erythroid progenitor cells, thymocytes, T cells, B cells and some tumor cells.

TRUSTED_SUSTAINABILITY

Specifications

Antigen IL-9R (CD129)
Applications Flow Cytometry
Classification Monoclonal
Clone AH9R7
Conjugate R718
Host Species Mouse
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Refrigerate Upon Arrival
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.