Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL10 Rabbit anti-Human, Unconjugated, Clone: IL10/2651R, Abnova™

Catalog No. 89337884 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-337-884 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-337-884 Supplier Abnova Corporation Supplier No. RAB00459
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody.

IL10 Monoclonal antibody specifically detects IL10 in Human samples. It is validated for IHC-P.

Specifications

Antigen IL10
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone IL10/2651R
Conjugate Unconjugated
Description interleukin 10
Gene IL10
Gene Alias CSIF, IL-10, IL10A, MGC126450, MGC126451, TGIF
Host Species Rabbit
Immunogen Original antibody is raised against recombinant protein corresponding to full length human IL10
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 3586
Target Species Human
Content And Storage Store at 2 to 8°C
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.