Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ IL10RA Monoclonal Antibody (C9), Invitrogen™
GREENER_CHOICE

Catalog No. PIMA543905
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA543905 100 μL
PIMA543906 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. PIMA543905 Supplier Invitrogen™ Supplier No. MA543905
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

The protein encoded by this gene is a receptor for interleukin 10. This protein is structurally related to interferon receptors. It has been shown to mediate the immunosuppressive signal of interleukin 10, and thus inhibits the synthesis of proinflammatory cytokines. This receptor is reported to promote survival of progenitor myeloid cells through the insulin receptor substrate-2/PI 3-kinase/AKT pathway. Activation of this receptor leads to tyrosine phosphorylation of JAK1 and TYK2 kinases. Two transcript variants, one protein-coding and the other not protein-coding, have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IL10RA
Applications Western Blot
Classification Monoclonal
Clone C9
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4
Gene Il10ra
Gene Accession No. Q13651
Gene Alias AW553859; CD210; CD210a; CDw210; CDW210A; CDW210B; CRF2-4; CRFB4; D21S58; D21S66; HIL-10R; I10R; I10R1; il-10 receptor; il-10 receptor antagonist; IL-10 receptor subunit alpha; IL10R; IL-10R alpha; IL-10R subunit 1; IL-10R subunit alpha; IL-10R1; IL-10R2; Il10ra; IL-10RA; interleukin 10 receptor subunit alpha; interleukin 10 receptor, alpha; interleukin-10 receptor alpha chain; interleukin-10 receptor subunit 1; Interleukin-10 receptor subunit alpha; mIL-10R
Gene Symbols Il10ra
Host Species Mouse
Immunogen Leu132-Asn235 (Accession Q13651), with N-terminal His Tag.
Purification Method Protein A/G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3587
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.