Learn More
IL10RA Mouse anti-Human, Clone: OTI1D10, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF506929

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
The protein encoded by this gene is a receptor for interleukin 10. This protein is structurally related to interferon receptors. It has been shown to mediate the immunosuppressive signal of interleukin 10, and thus inhibits the synthesis of proinflammatory cytokines. This receptor is reported to promote survival of progenitor myeloid cells through the insulin receptor substrate-2/PI 3-kinase/AKT pathway. Activation of this receptor leads to tyrosine phosphorylation of JAK1 and TYK2 kinases. Two transcript variants, one protein-coding and the other not protein-coding, have been found for this gene.Specifications
IL10RA | |
Monoclonal | |
Unconjugated | |
IL10RA | |
CD210, CD210a, CDW210A, HIL-10R, IL-10R1, IL10R | |
Mouse | |
Affinity Chromatography | |
RUO | |
3587 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Immunohistochemistry (Paraffin), Western Blot | |
OTI1D10 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q13651 | |
IL10RA | |
Full length human recombit protein of human IL10RA produced in HEK293T cell. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.