Learn More
Invitrogen™ IL10RA Polyclonal Antibody

Description
Recommended positive controls: HepG2, HCT116. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Specifications
Specifications
| Antigen | IL10RA |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 1.68 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 20% glycerol and 0.025% ProClin 300; pH 7 |
| Gene | Il10ra |
| Gene Accession No. | Q13651, Q61727 |
| Gene Alias | AW553859; CD210; CD210a; CDw210; CDW210A; CDW210B; CRF2-4; CRFB4; D21S58; D21S66; HIL-10R; I10R; I10R1; il-10 receptor; il-10 receptor antagonist; IL-10 receptor subunit alpha; IL10R; IL-10R alpha; IL-10R subunit 1; IL-10R subunit alpha; IL-10R1; IL-10R2; Il10ra; IL-10RA; interleukin 10 receptor subunit alpha; interleukin 10 receptor, alpha; interleukin-10 receptor alpha chain; interleukin-10 receptor subunit 1; Interleukin-10 receptor subunit alpha; mIL-10R |
| Gene Symbols | Il10ra |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.