Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ IL10RA Polyclonal Antibody

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA5109852

Catalog No. PIPA5109852


Only null left
Add to Cart

Description

Description

Immunogen sequence: KLPSVLLFKK PSPFIFISQR PSPETQDTIH PLDEEAFLKV SPELKNLDLH GSTDSGFGST KPSLQTEEPQ FLLPDPHPQA DRTLGNREPP VLGDSCSSGS SNSTDSGICL QEPSLSPSTG PTWEQQVGSN SRGQDDSGID LVQNSE.

The protein encoded by this gene is a receptor for interleukin 10. This protein is structurally related to interferon receptors. It has been shown to mediate the immunosuppressive signal of interleukin 10, and thus inhibits the synthesis of proinflammatory cytokines. This receptor is reported to promote survival of progenitor myeloid cells through the insulin receptor substrate-2/PI 3-kinase/AKT pathway. Activation of this receptor leads to tyrosine phosphorylation of JAK1 and TYK2 kinases. Two transcript variants, one protein-coding and the other not protein-coding, have been found for this gene.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

IL10RA
Polyclonal
Unconjugated
Il10ra
AW553859; CD210; CD210a; CDw210; CDW210A; CDW210B; CRF2-4; CRFB4; D21S58; D21S66; HIL-10R; I10R; I10R1; il-10 receptor; il-10 receptor antagonist; IL-10 receptor subunit alpha; IL10R; IL-10R alpha; IL-10R subunit 1; IL-10R subunit alpha; IL-10R1; IL-10R2; Il10ra; IL-10RA; interleukin 10 receptor subunit alpha; interleukin 10 receptor, alpha; interleukin-10 receptor alpha chain; interleukin-10 receptor subunit 1; Interleukin-10 receptor subunit alpha; mIL-10R
Rabbit
Affinity chromatography
RUO
117539, 16154, 3587
-20°C, Avoid Freeze/Thaw Cycles
Liquid
ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
0.31 mg/mL
PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3
Q13651, Q61727
Il10ra
Recombinant fusion protein containing a sequence corresponding to amino acids 265-410 of human IL10RA (NP_0015492).
100 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

WARNING: Cancer - www.P65Warnings.ca.gov
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.