Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IL10RB Monoclonal Antibody (01)
IL10RB Monoclonal Antibody (1)
Supplier: Thermo Scientific MA530524
Description

Specifications
IL10RB | |
Monoclonal | |
1 mg/mL | |
PBS with no preservative | |
Q08334 | |
IL10RB | |
Recombinant Human IL10RB/IL-10RB Protein (Met1-Ser220). | |
100 μL | |
Primary | |
Human | |
Antibody |
ELISA | |
01 | |
Unconjugated | |
IL10RB | |
6620401D04Rik; AI528744; CDW210B; CRF2-4; CRFB4; cytokine receptor class-II CRF2-4; cytokine receptor class-II member 4; Cytokine receptor family 2 member 4; cytokine receptor family II, member 4; D16H21S58; D21S58; D21S58h; D21S66; IL-10 receptor subunit beta; IL-10R subunit 2; IL-10R subunit beta; Il10r2; IL-10R2; IL10RB; IL-10RB; inte; interleukin 10 receptor 2; interleukin 10 receptor subunit beta; interleukin 10 receptor, beta; interleukin-10 receptor subunit 2; Interleukin-10 receptor subunit beta; RGD1560373 | |
Mouse | |
Protein A | |
RUO | |
3588 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction