Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ IL10RB Monoclonal Antibody (1D12C12)
GREENER_CHOICE

Catalog No. FERMA548510
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FERMA548510 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FERMA548510 Supplier Invitrogen™ Supplier No. MA548510
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

IL10RB Monoclonal Antibody for Western Blot, ELISA

Natural killer (NK) cells in teleosts and their evolutionary homologue are a subpopulation of lymphocytes with properties that distinguish them from either B- or T-cells. NK cells are important effectors of innate immunity where they release cytokines, which in turn up-regulate other immunological functions. Monoclonal antibodies have been used to identify different surface antigens present on NK cells. These surface antigens have not only been used to identify NK cells, but also their functionally distinct subsets. The Cluster of Differentiation (CD) nomenclature was established to standardize the naming of NK cells.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IL10RB
Applications ELISA, Western Blot, Immunocytochemistry, Immunohistochemistry, Western Blot
Classification Monoclonal
Clone 1D12C12
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% sodium azide
Gene IL10RB
Gene Accession No. Q08334
Gene Alias 6620401D04Rik; AI528744; CDW210B; CRF2-4; CRFB4; cytokine receptor class-II CRF2-4; cytokine receptor class-II member 4; Cytokine receptor family 2 member 4; cytokine receptor family II, member 4; D16H21S58; D21S58; D21S58h; D21S66; IL-10 receptor subunit beta; IL-10R subunit 2; IL-10R subunit beta; Il10r2; IL-10R2; IL10RB; IL-10RB; inte; interleukin 10 receptor 2; interleukin 10 receptor subunit beta; interleukin 10 receptor, beta; interleukin-10 receptor subunit 2; Interleukin-10 receptor subunit beta; RGD1560373
Gene Symbols IL10RB
Host Species Mouse
Immunogen Purified recombinant fragment of human IL10RB (AA: extra 20-220) expressed in E. Coli.
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3588
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.